• USA Home
  • 45415 - Erbium(III) trifluoromethanesulfonate

EMAIL THIS PAGE TO A FRIEND
45415 Sigma-Aldrich

Erbium(III) trifluoromethanesulfonate

purum, ≥97.0% (KT)

Synonym: Erbium(III) triflate, Trifluoromethanesulfonic acid erbium(III) salt

Purchase

Properties

InChI Key   GLQOFBCJADYRKR-UHFFFAOYSA-K
grade   purum
assay   ≥97.0% (KT)

Description

Other Notes

Lewis acid catalyst which can be used in aqueous solution and easily recycled. Employed in aldol reactions of silyl enol ethers with aldehydes; Triflate-catalyzed asymmetric Diels-Alder reaction

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

425672 Erbium(III) trifluoromethanesulfonate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?