EMAIL THIS PAGE TO A FRIEND
45419 Sigma-Aldrich

Dazomet

PESTANAL®, analytical standard

Synonym: 3,5-Dimethyltetrahydro-1,3,5-thiadiazine-2-thione, Thiadiazine

  • CAS Number 533-74-4

  • Empirical Formula (Hill Notation) C5H10N2S2

  • Molecular Weight 162.28

  •  Beilstein Registry Number 116039

  •  EC Number 208-576-7

  •  MDL number MFCD00023809

  •  PubChem Substance ID 24868897

Purchase

Properties

Related Categories All Fungicides, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, D,
grade   analytical standard
InChI Key   QAYICIQNSGETAS-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
XI2800000
Flash Point(F) 
312.8 °F
Flash Point(C) 
156 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?