EMAIL THIS PAGE TO A FRIEND
45420 Sigma-Aldrich

2,4-DB

PESTANAL®, analytical standard

Synonym: 2,4-DB, 4-(2,4-Dichlorophenoxy)butyric acid

  • CAS Number 94-82-6

  • Linear Formula Cl2C6H3O(CH2)3CO2H

  • Molecular Weight 249.09

  •  Beilstein Registry Number 1976809

  •  EC Number 202-366-9

  •  MDL number MFCD00002819

  •  PubChem Substance ID 24868898

Purchase

Properties

Related Categories A-D, All Herbicide Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   YIVXMZJTEQBPQO-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   118-120 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
ES9100000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?