EMAIL THIS PAGE TO A FRIEND
45440 Sigma-Aldrich

1,3-Dichloropropene

PESTANAL®, analytical standard

  • CAS Number 542-75-6

  • Linear Formula ClCH2CH=CHCl

  • Molecular Weight 110.97

  •  Beilstein Registry Number 1719556

  •  EC Number 208-826-5

  •  MDL number MFCD00000985

  •  PubChem Substance ID 329756735

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, D, Nematicides,
grade   analytical standard
description   cis+trans
  mixture of isomers
InChI Key   UOORRWUZONOOLO-OWOJBTEDSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.472(lit.)
bp   97-112 °C(lit.)
density   1.198 g/mL at 25 °C(lit.)
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN 1992 6.1(3) / PGIII
WGK Germany 
3
RTECS 
UC8310000
Flash Point(F) 
82.4 °F
Flash Point(C) 
28 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?