EMAIL THIS PAGE TO A FRIEND
45457 Sigma-Aldrich

Diphenylmercury(II)

PESTANAL®, analytical standard

Synonym: Mercury(II)diphenyl

  • CAS Number 587-85-9

  • Linear Formula (C6H5)2Hg

  • Molecular Weight 354.80

  •  Beilstein Registry Number 637234

  •  EC Number 209-606-1

  •  MDL number MFCD00002998

  •  PubChem Substance ID 24868936

Purchase

Properties

Related Categories Additional Standards, Alpha Index of All Pesticide Standards, Amines, Analytical Standards, Analytical/Chromatography,
grade   PESTANAL®, analytical standard
InChI Key   HWMTUNCVVYPZHZ-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   121-124 °C(lit.)
density   2.32 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2026 6.1 / PGIII
WGK Germany 
3
RTECS 
OW3150000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?