EMAIL THIS PAGE TO A FRIEND
45498 Sigma-Aldrich

Flamprop-methyl

PESTANAL®, analytical standard

Purchase

Properties

Related Categories All Herbicide Standards, Alpha Index of All Pesticide Standards, Amide, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   RBNIGDFIUWJJEV-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
UE8900000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?