EMAIL THIS PAGE TO A FRIEND
45500 Sigma-Aldrich

Fluchloralin

PESTANAL®, analytical standard

  • CAS Number 33245-39-5

  • Empirical Formula (Hill Notation) C12H13ClF3N3O4

  • Molecular Weight 355.70

  •  Beilstein Registry Number 2778716

  •  EC Number 251-426-0

  •  MDL number MFCD00078686

  •  PubChem Substance ID 24869006

Purchase

Properties

Related Categories All Herbicide Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Dinitroaniline,
grade   analytical standard
InChI Key   MNFMIVVPXOGUMX-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
XU5160000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?