• USA Home
  • 45512 - Ergotamine D-tartrate

EMAIL THIS PAGE TO A FRIEND
45512 Sigma-Aldrich

Ergotamine D-tartrate

tested according to Ph.Eur.

Synonym: Ergotamini tartras

  • CAS Number 379-79-3

  • Empirical Formula (Hill Notation) C33H35N5O5 · 0.5C4H6O6

  • Molecular Weight 656.70

  •  Beilstein Registry Number 6173437

  •  EC Number 206-835-9

  •  MDL number MFCD08277639

  •  PubChem Substance ID 329756799

Purchase

Properties

Related Categories Analytical/Chromatography, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Heterocyclic Building Blocks,
InChI Key   CJMJLDQKTOJACI-BGQAIRJTSA-N
pharmacopeia   testing conforms to Pharmacopeia
mp   ~195 °C (dec.)(lit.)
storage temp.   2-8°C

Description

Application

Ergotamine D-tartrate may be used as a standard during the quantitative estimation of ergot alkaloids in vein tissue by liquid chromatography-mass spectrometry (LC-MS).

General description

Ergotamine D-tartrate is an ergot alkaloid.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1544PSN2 6.1 / PGI
WGK Germany 
3
RTECS 
KE8225000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Carcinogen Selector - Digestive System - Small Intestine

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Small Intestine
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System - Stomach

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Stomach
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?