• USA Home
  • 45525 - Hexachloro-1,3-butadiene

EMAIL THIS PAGE TO A FRIEND
45525 Sigma-Aldrich

Hexachloro-1,3-butadiene

PESTANAL®, analytical standard

Synonym: Perchlorobutadiene

  • CAS Number 87-68-3

  • Linear Formula CCl2=CClCCl=CCl2

  • Molecular Weight 260.76

  •  Beilstein Registry Number 1766570

  •  EC Number 201-765-5

  •  MDL number MFCD00000836

  •  PubChem Substance ID 24869044

Purchase

Properties

Related Categories All Fungicides, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Fungicides,
grade   analytical standard
vapor pressure   0.2 mmHg ( 20 °C)
InChI Key   RWNKSTSCBHKHTB-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.555(lit.)
bp   210-220 °C(lit.)
mp   −22-−19 °C(lit.)
density   1.665 g/mL at 25 °C(lit.)
format   neat
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2279 6.1 / PGIII
WGK Germany 
3
RTECS 
EJ0700000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?