EMAIL THIS PAGE TO A FRIEND

45550 Sigma-Aldrich

Calcon

indicator (for complexometry (Al, Fe, Zr))

Synonym: 1-(2-Hydroxy-1-naphthylazo)-2-naphthol-4-sulfonic acid sodium salt, Eriochrome® Blue Black R, Mordant Black 17, Palatine Chrome Black 6BN

  • CAS Number 2538-85-4

  • Empirical Formula (Hill Notation) C20H13N2NaO5S

  • Molecular Weight 416.38

  •  Beilstein Registry Number 6041435

  •  Colour Index Number 15705

  •  EC Number 219-810-2

  •  MDL number MFCD00004069

  •  PubChem Substance ID 24869074

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Indicators, Metal Titration Indicators, Titration More...
grade   indicator (for complexometry (Al, Fe, Zr))

Description

General description

Visit our Titration Center to learn more.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Legal Information

Calcon is a trademark of Benckiser

Eriochrome is a registered trademark of Huntsman Petrochemical, LLC

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Explore the New Analytical Standards Portal

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
QK2200000

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

[Recent advances on reaction mechanism of anthocyanin biosynthesis]. Nakajima J, Yamazaki M, and Saito K Tanpakushitsu Kakusan Koso. 47(3), 217-24, (2002)

Loading...


Removal of Eriochrome Blue Black R from wastewater using micellar-enhanced ultrafiltration. Zaghbani N, Hafiane A, and Dhahbi M J. Hazard. Mater. 168(2-3), 1417-21, (2009)

Loading...


The effect of pH in the adsorption of Alizarin and Eriochrome Blue Black R onto iron oxides. Pirillo S, Ferreira ML, and Rueda EH J. Hazard. Mater. 168(1), 168-78, (2009)

Loading...


Fluorimetric assay of aluminum in tablets by pontachrome blue black reagent. Raggi MA, Varani G, Nobile L, et al. Pharmazie 42(2), 134-5, (1987)

Loading...


SAXS and ATR-FTIR studies on EBT-TSX mixtures in their sol-gel phases. Hirun N, Rugmai S, Sangfai T, et al. Int. J. Biol. Macromol. 51(4), 423-30, (2012)

Loading...


[Spectra studies on interaction of bovine serum albumin with acidic chrome blue K in acidic solution]. Yu Y, Liao J, and Huang FD Guang Pu Xue Yu Guang Pu Fen Xi 22(6), 1067-9, (2002)

Loading...


Resonance light scattering study on the interaction of benproperine phosphate with eriochrome blue black R in the presence of sodium dodecylbenzene sulphonate and its analytical application. Feng S, Liu J, Han X, et al. Luminescence 24(2), 67-72, (2009)

Loading...


[A catalytic kinetic spectrophotometric method for determination of trace zinc(II) in Mongolian medicine and hair sample]. Sha R, Bao D, Wu D, et al. Guang Pu Xue Yu Guang Pu Fen Xi 24(12), 1646-8, (2004)

Loading...


[Colorimetric and fluorometric determinations of cartap, an insecticide, by ion association with calcon]. Sano T and Syoyama M Yakugaku Zasshi 103(6), 650-4, (1983)

Loading...


Beil. 16,IV,428

FT-IR 1 (2), 987:A / FT-IR 2 (3), 4094:A / IR-Spectra (3), 1462:F / RegBook 1 (2), 2751:B / Sax 6, 624 / Sigma FT-IR 1 (2), 151:D / Stains and Dyes Ref, 545 / Structure Index 1, 435:B:7

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?