EMAIL THIS PAGE TO A FRIEND
45635 Sigma-Aldrich

Prometon

PESTANAL®, analytical standard

Synonym: 2,4-Bis-(isopropylamino)-6-methoxy-1,3,5-triazine

  • CAS Number 1610-18-0

  • Empirical Formula (Hill Notation) C10H19N5O

  • Molecular Weight 225.29

  •  Beilstein Registry Number 613574

  •  EC Number 216-548-0

  •  MDL number MFCD00047343

  •  PubChem Substance ID 24869164

Purchase

Properties

Related Categories Additional Standards, All Herbicide Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   ISEUFVQQFVOBCY-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
XY4200000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?