EMAIL THIS PAGE TO A FRIEND
45652 Sigma-Aldrich

Quinonamid

PESTANAL®, analytical standard

  • CAS Number 27541-88-4

  • Empirical Formula (Hill Notation) C12H6Cl3NO3

  • Molecular Weight 318.54

  •  Beilstein Registry Number 2146844

  •  EC Number 248-516-7

  •  MDL number MFCD00055482

  •  PubChem Substance ID 329756866

Purchase

Properties

Related Categories Algicides, Alpha Index of All Pesticide Standards, Amide, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   ZIEWAMOXCOLNSJ-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?