EMAIL THIS PAGE TO A FRIEND
45654 Sigma-Aldrich

Rabenzazol

PESTANAL®, analytical standard

Synonym: 1-Benzimidazolyl-3,5-dimethylpyrazole

  • CAS Number 40341-04-6

  • Empirical Formula (Hill Notation) C12H12N4

  • Molecular Weight 212.25

  •  Beilstein Registry Number 959559

  •  EC Number 254-882-9

  •  MDL number MFCD00055495

  •  PubChem Substance ID 329756868

Purchase

Properties

Related Categories All Fungicides, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Benzimidazoles,
grade   analytical standard
InChI Key   RUGYNGIMTAFTLP-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DD8780000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?