EMAIL THIS PAGE TO A FRIEND
45663 Sigma-Aldrich

Sulfallate

PESTANAL®, analytical standard

  • CAS Number 95-06-7

  • Empirical Formula (Hill Notation) C8H14ClNS2

  • Molecular Weight 223.79

  •  Beilstein Registry Number 1772271

  •  EC Number 202-388-9

  •  MDL number MFCD00048530

  •  PubChem Substance ID 329756877

Purchase

Properties

Related Categories All Herbicide Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Herbicides,
grade   PESTANAL®, analytical standard
InChI Key   XJCLWVXTCRQIDI-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
EZ5075000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Carcinogen Selector - Digestive System - Stomach

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Stomach
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Mammary Gland

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System - Mammary Gland
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Lung

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Reproductive System - Lung
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Respiratory System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Respiratory System
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?