EMAIL THIS PAGE TO A FRIEND
45669 Sigma-Aldrich

TBTO

PESTANAL®, analytical standard

Synonym: Bis[tri-n-butyltin(IV)]oxide, HBD, Hexabutyldistannoxane, TBTO, Tributyltin(IV) oxide

  • CAS Number 56-35-9

  • Linear Formula (CH3CH2CH2CH2)3SnOSn(CH2CH2CH2CH3)3

  • Molecular Weight 596.10

  •  Beilstein Registry Number 745057

  •  EC Number 200-268-0

  •  MDL number MFCD00009418

  •  PubChem Substance ID 24869220

Purchase

Properties

Related Categories 2000/60/EC, Additional Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
vapor pressure   <0.01 mmHg ( 25 °C)
InChI Key   APQHKWPGGHMYKJ-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.486(lit.)
bp   180 °C/2 mmHg(lit.)
density   1.17 g/mL at 25 °C(lit.)
format   neat
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2788 6.1 / PGII
WGK Germany 
3
RTECS 
JN8750000
Flash Point(F) 
374 °F
Flash Point(C) 
190 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?