• USA Home
  • 45722 - Dichloroprop-2-ethylhexyl ester

EMAIL THIS PAGE TO A FRIEND
45722 Sigma-Aldrich

Dichloroprop-2-ethylhexyl ester

PESTANAL®, analytical standard

Synonym: 2-Ethylhexyl 2-(2,4-dichlorophenoxy)propionate

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, D, Pesticides & Metabolites Standards More...
grade   PESTANAL®, analytical standard
InChI Key   CEEDFYRUPAWDOU-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?