• USA Home
  • 45785 - 1,6-Dimethylnaphthalene

EMAIL THIS PAGE TO A FRIEND
45785 Sigma-Aldrich

1,6-Dimethylnaphthalene

analytical standard

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
InChI Key   CBMXCNPQDUJNHT-UHFFFAOYSA-N
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.606(lit.)
bp   265-266 °C(lit.)
mp   −17-−16 °C(lit.)
density   1.002 g/mL at 25 °C(lit.)
format   neat
Gene Information   human ... CYP1A2(1544)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
233.6 °F
Flash Point(C) 
112 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?