• USA Home
  • 45786 - 1,8-Dimethylnaphthalene

EMAIL THIS PAGE TO A FRIEND
45786 Sigma-Aldrich

1,8-Dimethylnaphthalene

analytical standard

  • CAS Number 569-41-5

  • Linear Formula C10H6(CH3)2

  • Molecular Weight 156.22

  •  Beilstein Registry Number 2039841

  •  EC Number 209-314-4

  •  MDL number MFCD00004040

  •  PubChem Substance ID 24869401

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
InChI Key   XAABPYINPXYOLM-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   270 °C(lit.)
mp   59-61 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?