EMAIL THIS PAGE TO A FRIEND
45791 Sigma-Aldrich

Indene

analytical standard

  • CAS Number 95-13-6

  • Empirical Formula (Hill Notation) C9H8

  • Molecular Weight 116.16

  •  Beilstein Registry Number 635873

  •  EC Number 202-393-6

  •  MDL number MFCD00003777

  •  PubChem Substance ID 329756954

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
InChI Key   YBYIRNPNPLQARY-UHFFFAOYSA-N
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.595(lit.)
bp   181-182 °C(lit.)
mp   −5-−3 °C(lit.)
density   0.996 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3295C 3 / PGIII
WGK Germany 
1
RTECS 
NK8225000
Flash Point(F) 
136.4 °F
Flash Point(C) 
58 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?