EMAIL THIS PAGE TO A FRIEND
45797 Sigma-Aldrich

Pentacene

analytical standard

  • CAS Number 135-48-8

  • Empirical Formula (Hill Notation) C22H14

  • Molecular Weight 278.35

  •  Beilstein Registry Number 1912418

  •  EC Number 205-193-7

  •  MDL number MFCD00003710

  •  PubChem Substance ID 329756958

Purchase

Properties

Related Categories Additional Standards, All Polycyclic Aromatic Hydrocarbon (PAH) Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   SLIUAWYAILUBJU-UHFFFAOYSA-N
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   372-374 °C (subl.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?