EMAIL THIS PAGE TO A FRIEND
45812 Sigma-Aldrich

DCU

PESTANAL®, analytical standard

Synonym: 1,3-Bis(1-hydroxy-2,2,2-trichloroethyl)urea, Dichlorolurea

  • CAS Number 116-52-9

  • Empirical Formula (Hill Notation) C5H6Cl6N2O3

  • Molecular Weight 354.83

  •  Beilstein Registry Number 1813364

  •  EC Number 204-144-7

  •  MDL number MFCD00018836

  •  PubChem Substance ID 329756968

Purchase

Properties

Related Categories A-D, All Herbicide Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   PPJXIHLNYDVTDI-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
YS2787000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?