• USA Home
  • 45844 - Dichlofluanid solution

EMAIL THIS PAGE TO A FRIEND
45844 Sigma-Aldrich

Dichlofluanid solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

  • CAS Number 1085-98-9

  • Empirical Formula (Hill Notation) C9H11Cl2FN2O2S2

  • Molecular Weight 333.23

  •  Beilstein Registry Number 2947992

  •  EC Number 200-835-2

  •  MDL number MFCD00078639

  •  PubChem Substance ID 329756984

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Amide structure, Analytical Standards, Analytical/Chromatography, D,
grade   analytical standard
InChI Key   WURGXGVFSMYFCG-UHFFFAOYSA-N
product line   PESTANAL®
shelf life   limited shelf life, expiry date on the label
concentration   100 μg/mL in acetonitrile
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 5) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 5) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

GC Analysis of Agricultural Pesticides (Standard) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agricultural Pesticides (Standard) on SLB®-5ms
Keywords: Agriculture, Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Herbicides, Mass spectrometry, Pesticides

GC Analysis of Agriculture Pesticides in Orange (Spiked) on SLB®-5ms after QuEChERS Cleanup using Primary-Secondary Amine (PSA)

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agriculture Pesticides in Orange (Spiked) on SLB®-5ms after QuEChERS Cleanup using Primary-Secondary Amine (PSA)
Keywords: Chromatography, Gas chromatography, Pesticides, Purification, Sample preparations, Solid phase extractions

GC Analysis of Pesticides in Wine on SLB®-5ms after SPME using 85 μm Polyacrylate Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Pesticides in Wine on SLB®-5ms after SPME using 85 μm Polyacrylate Fiber
Keywords: Chromatography, Gas chromatography, Metabolites, Pesticides, Purification, Solid phase microextractions

GC Analysis of a 19-Component Pesticide Mix on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 19-Component Pesticide Mix on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Pesticides, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?