EMAIL THIS PAGE TO A FRIEND
45853 Sigma-Aldrich

(+)-Bornyl acetate

analytical standard

Synonym: (+)-Acetic acid bornyl ester, endo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]hept-2-yl acetate

  • CAS Number 20347-65-3

  • Empirical Formula (Hill Notation) C12H20O2

  • Molecular Weight 196.29

  •  Beilstein Registry Number 1909308

  •  EC Number 243-750-6

  •  MDL number MFCD00135942

  •  PubChem Substance ID 24869473

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Asymmetric Synthesis, Chemical Class,
grade   analytical standard
InChI Key   KGEKLUUHTZCSIP-SCVCMEIPSA-N
assay   ≥98.5% (sum of enantiomers, GC)
form   neat
optical activity   [α]20/D +42±1°, neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.463
density   0.984 g/mL at 20 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NA 1993 / PGIII
WGK Germany 
1
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?