458600 Sigma-Aldrich

Ethanol

denatured, (UK IDA standard)

DOWNLOAD MSDS (PDF)

Synonym: Ethyl alcohol UK Industrial methylated spirit

  • CAS Number 64-17-5

  • Linear Formula CH3CH2OH

  • Molecular Weight 46.07

  •  Beilstein Registry Number 1718733

  •  MDL number MFCD00003568

  •  PubChem Substance ID 24869487

Properties

grade   denatured
vapor density   1.59 (vs air)
vapor pressure   44.6 mmHg ( 20 °C)
autoignition temp.   683 °F
expl. lim.   19 %, 60 °F
availability   not available in Switzerland
refractive index   n20/D 1.3600(lit.)
bp   78 °C(lit.)
mp   −114 °C(lit.)
density   0.789 g/mL at 25 °C(lit.)

Description

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Price and Availability

We Have the Silica You Need and a New Brochure

Silica Gel PurificationWhether you're looking for a specific brand, quality, or price, Sigma-Aldrich has the silica products to meet your unique purification or sample preparation requirements. Our new Silica Brochure makes it easy to find just the silica gel product you need.


Learn more.

Customers Also Purchased

24201Acetone

puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)

CHROMASOLV®, for HPLC, ≥99.8%, contains amylene as stabilizer

CHROMASOLV®, for HPLC, ≥99.7%

34873Heptane

CHROMASOLV®, for HPLC, ≥99%

CHROMASOLV®, for HPLC, ≥99.9%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?