EMAIL THIS PAGE TO A FRIEND
45864 Sigma-Aldrich

Hexazinone solution

PESTANAL®, analytical standard, 100 ng/μL in methanol

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, H-M, Herbicides,
grade   analytical standard
InChI Key   CAWXEEYDBZRFPE-UHFFFAOYSA-N
product line   PESTANAL®
concentration   100 ng/μL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Nitrogen-Containing Herbicides in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Nitrogen-Containing Herbicides in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

HPLC Analysis of Atrazine Herbicides on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Atrazine Herbicides on Ascentis® Express C18
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

36129 Hexazinone, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?