EMAIL THIS PAGE TO A FRIEND
45867 Sigma-Aldrich

Isoproturon solution

100 ng/μL in acetonitrile, PESTANAL®, analytical standard

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Food and Cosmetic Component Standards, H-M,
grade   PESTANAL®, analytical standard
InChI Key   PUIYMUZLKQOUOZ-UHFFFAOYSA-N
concentration   100 ng/μL in acetonitrile
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of Atrazine Herbicides on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Atrazine Herbicides on Ascentis® Express C18
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of Phenyl Urea Pesticides on Ascentis® Express C18 Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Phenyl Urea Pesticides on Ascentis® Express C18 Phase
Keywords: Chromatography, Herbicides, High performance liquid chromatography, Pesticides, Separation, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

36137 Isoproturon, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?