EMAIL THIS PAGE TO A FRIEND
45868 Sigma-Aldrich

Linuron solution

100 μg/mL in methanol, PESTANAL®, analytical standard

  • CAS Number 330-55-2

  • Empirical Formula (Hill Notation) C9H10Cl2N2O2

  • Molecular Weight 249.09

  •  Beilstein Registry Number 2128725

  •  EC Number 200-659-6

  •  MDL number MFCD00055249

  •  PubChem Substance ID 329756997

Purchase

Properties

Related Categories 2000/60/EC, Additional Standards, All Herbicide Standards, Alpha Index of All Pesticide Standards, Analytical Standards,
grade   analytical standard
InChI Key   XKJMBINCVNINCA-UHFFFAOYSA-N
product line   PESTANAL®
shelf life   limited shelf life, expiry date on the label
concentration   100 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles

Articles

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of Atrazine Herbicides on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Atrazine Herbicides on Ascentis® Express C18
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of Carbamate and Urea Pesticides on SUPELCOSIL™ LC-8 after SPME using 60 μm PDMS/DVB Fiber

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Carbamate and Urea Pesticides on SUPELCOSIL™ LC-8 after SPME using 60 μm PDMS/DVB Fiber
Keywords: Chromatography, Environmental, Environmental Analysis, High performance liquid chromatography, Metabolites, Pesticides, Solid phase microextractions

HPLC Analysis of Carbamate and Urea Pesticides on SUPELCOSIL™ LC-8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Carbamate and Urea Pesticides on SUPELCOSIL™ LC-8
Keywords: Chromatography, Environmental, Environmental Analysis, Herbicides, High performance liquid chromatography, Pesticides

HPLC Analysis of Nonvolatile Pesticides in Water on SUPELCOSIL™ LC-18-DB after SPE using Supelclean™ ENVI-Carb™

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Nonvolatile Pesticides in Water on SUPELCOSIL™ LC-18-DB after SPE using Supelclean™ ENVI-Carb™
Keywords: Chromatography, Environmental, Environmental Analysis, High performance liquid chromatography, Metabolites, Pesticides, Sample preparations, Solid phase extractions

HPLC Analysis of Phenyl Urea Pesticides on Ascentis® Express C18 Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Phenyl Urea Pesticides on Ascentis® Express C18 Phase
Keywords: Chromatography, Herbicides, High performance liquid chromatography, Pesticides, Separation, Size-exclusion chromatography

HPLC Analysis of Urea Pesticides on Discovery® Cyano

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Urea Pesticides on Discovery® Cyano
Keywords: Chromatography, Environmental, Environmental Analysis, Herbicides, High performance liquid chromatography, Metabolites, Pesticides

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?