EMAIL THIS PAGE TO A FRIEND
45873 Sigma-Aldrich

MCPA solution

100 ng/μL in acetonitrile, PESTANAL®, analytical standard

Synonym: (4-Chloro-2-methylphenoxy)acetic acid solution

  • CAS Number 94-74-6

  • Empirical Formula (Hill Notation) C9H9ClO3

  • Molecular Weight 200.62

  •  Beilstein Registry Number 2051752

  •  EC Number 200-835-2

  •  MDL number MFCD00004306

  •  PubChem Substance ID 329756999

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, H-M, Herbicides,
grade   analytical standard
InChI Key   WHKUVVPPKQRRBV-UHFFFAOYSA-N
product line   PESTANAL®
shelf life   limited shelf life, expiry date on the label
concentration   100 ng/μL in acetonitrile
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?