• USA Home
  • 45907 - 2,3,4,6-Tetrachlorophenol solution

EMAIL THIS PAGE TO A FRIEND
45907 Sigma-Aldrich

2,3,4,6-Tetrachlorophenol solution

100 μg/mL in methanol, PESTANAL®, analytical standard

  • CAS Number 58-90-2

  • Empirical Formula (Hill Notation) C6H2Cl4O

  • Molecular Weight 231.89

  •  Beilstein Registry Number 779754

  •  EC Number 200-659-6

  •  MDL number MFCD00019979

  •  PubChem Substance ID 329757010

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Pesticides & Metabolites Standards, Q-Z More...
grade   analytical standard
InChI Key   VGVRPFIJEJYOFN-UHFFFAOYSA-N
product line   PESTANAL®
shelf life   limited shelf life, expiry date on the label
concentration   100 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles

Articles

GC Analysis of Phenols in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI-Chrom P

From our library of Articles, Sigma-Aldrich presents GC Analysis of Phenols in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI-Chrom P
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Sample preparations, Size-exclusion chromatography, Solid phase extractions

GC Analysis of Phenols in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Phenols in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Phenols on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Phenols on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Trichloroanisole (TCA) and Precursors in Red Wine (Spiked) on SLB-5ms after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Trichloroanisole (TCA) and Precursors in Red Wine (Spiked) on SLB-5ms after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Electron capture detector, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 8041: GC Analysis of Phenols on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8041: GC Analysis of Phenols on Equity®-5
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?