• USA Home
  • 45926 - 1-Chloro-2-nitrobenzene

EMAIL THIS PAGE TO A FRIEND
45926 Sigma-Aldrich

1-Chloro-2-nitrobenzene

analytical standard

  • CAS Number 88-73-3

  • Linear Formula ClC6H4NO2

  • Molecular Weight 157.55

  •  Beilstein Registry Number 509273

  •  EC Number 201-854-9

  •  MDL number MFCD00007061

  •  PubChem Substance ID 329757015

Purchase

Properties

Related Categories A-C, Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
vapor density   5.4 (vs air)
vapor pressure   0.04 mmHg ( 25 °C)
InChI Key   BFCFYVKQTRLZHA-UHFFFAOYSA-N
form   neat
autoignition temp.   500 °F
shelf life   limited shelf life, expiry date on the label
expl. lim.   8.7 %
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   246 °C(lit.)
mp   31-33 °C(lit.)
density   1.348 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1578 6.1 / PGII
WGK Germany 
3
RTECS 
CZ0875000
Flash Point(F) 
258.8 °F
Flash Point(C) 
126 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?