• USA Home
  • 45929 - 4-Methyl-3-nitroaniline

EMAIL THIS PAGE TO A FRIEND
45929 Sigma-Aldrich

4-Methyl-3-nitroaniline

analytical standard

Synonym: 3-Nitro-4-methylaniline, 3-Nitro-p-toluidine, 4-Amino-2-nitro-toluene

  • CAS Number 119-32-4

  • Linear Formula CH3C6H3(NO2)NH2

  • Molecular Weight 152.15

  •  Beilstein Registry Number 638984

  •  EC Number 204-314-0

  •  MDL number MFCD00007910

  •  PubChem Substance ID 329757016

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Amines, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   GDIIPKWHAQGCJF-UHFFFAOYSA-N
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   74-77 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2660 6.1 / PGIII
WGK Germany 
3
RTECS 
XU8227000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?