• USA Home
  • 45946 - 4-Amino-2-nitrophenol

EMAIL THIS PAGE TO A FRIEND
45946 Sigma-Aldrich

4-Amino-2-nitrophenol

analytical standard

Synonym: 4-Hydroxy-3-nitroaniline

  • CAS Number 119-34-6

  • Linear Formula H2NC6H3(NO2)OH

  • Molecular Weight 154.12

  •  Beilstein Registry Number 1368435

  •  EC Number 204-316-1

  •  MDL number MFCD00007876

  •  PubChem Substance ID 329757020

Purchase

Properties

Related Categories A-B, Additional Standards, Alpha Sort, Amines, Analytical Standards,
grade   analytical standard
InChI Key   WHODQVWERNSQEO-UHFFFAOYSA-N
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   125-127 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SJ6303000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?