• USA Home
  • 45957 - 1-Chloro-3-nitrobenzene

EMAIL THIS PAGE TO A FRIEND
45957 Sigma-Aldrich

1-Chloro-3-nitrobenzene

analytical standard

Purchase

Properties

Related Categories A-C, Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   KMAQZIILEGKYQZ-UHFFFAOYSA-N
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   236 °C(lit.)
mp   43-47 °C(lit.)
density   1.534 g/mL at 25 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 1578 6.1 / PGII
WGK Germany 
3
RTECS 
CZ0940000
Flash Point(F) 
239 °F
Flash Point(C) 
115 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?