EMAIL THIS PAGE TO A FRIEND
45967 Sigma-Aldrich

1,4-Dinitrobenzene

analytical standard

  • CAS Number 100-25-4

  • Linear Formula C6H4(NO2)2

  • Molecular Weight 168.11

  •  Beilstein Registry Number 1105828

  •  EC Number 202-833-7

  •  MDL number MFCD00007314

  •  PubChem Substance ID 24869582

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
InChI Key   FYFDQJRXFWGIBS-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   183.4 °C/34 mmHg(lit.)
mp   170-173 °C(lit.)
density   1.625 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3443 6.1 / PGII
WGK Germany 
3
RTECS 
CZ7525000
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?