EMAIL THIS PAGE TO A FRIEND
459828 Sigma-Aldrich

Ethyl alcohol, Pure

200 proof, HPLC/spectrophotometric grade

Synonym: Ethanol, absolute alcohol, non-denatured ethanol

Purchase

Properties

Related Categories Amber Glass Bottles, Ethanol (Ethyl Alcohol), Ethanol (Tax-Paid), HPLC and Spectrophotometric, HPLC, HPLC Plus, and HPLC Gradient Grades,
grade   HPLC/spectrophotometric grade
assay   ≥99.5%
availability   not available in (Sales restictions may apply)
concentration   200 proof
impurities   ≤0.2% water
evapn. residue   <0.001%
color   colorless
bp   78.3 °C
mp   -114 °C
density   0.789 g/mL at 20 °C
λ   H2O reference
UV absorption   λ: 210 nm Amax: 0.40
  λ: 220 nm Amax: 0.25
  λ: 230 nm Amax: 0.15
  λ: 240 nm Amax: 0.05
  λ: 270-400 nm Amax: 0.01

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pure-Pac® II containers require the Micromatic MicroValve coupler for dispensing solvents, Z560723.

Sales restrictions may apply

Packaging

1, 6×1, 2, 4, 4×4 L in glass bottle

20 L in Nowpak™

View returnable container options.

20 L in Nowpak™ II

Application

Pure ethyl alcohol (Ethanol) has been used to prepare:
• Ethanol solution for the synthesis of Ga-68 DOTATATE [DOTA-(Tyr3)-octreotate (where DOTA = 1,4,7,10-tetraazacyclo- dodecane-1,4,7,10-tetraacetic acid)] by an automated synthesis system.
• Aristolochic acid stock solution for the spectrophotometric analysis.
• Core silica particles of nanoshells by a modified Stöber method.
• Polyvinylpyrrolidone (PVP) solution in combination with water for synthesizing PVP ultrafine fibers by solution (air-) blowing method.

General description

Price includes Federal Tax.

Pure ethyl alcohol is a non-denatured 200proof ethanol. Ethanol, a primary alcohol, is an important raw material for the synthesis of various compounds. It can be synthesized either from molasses by fermentation or by hydration of ethylene. It acts as a hydrogen source and reaction medium during the transfer hydrogenation of carbonyl compounds in the presence of sodium hydroxide. The impact of ethanol/water fumigation on the performance of diesel engines has been studied. Spectrophotometric grade solvent is of high purity with low UV absorption.

Legal Information

Pure-Pac is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol
WGK Germany 
1
RTECS 
KQ6300000
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

Z568031 Pure Solv Micro 100 Liter solvent purification system, with activated Pure Solv media column, joint Joint: ST/NS 24/40 US
Z568074 Pure Solv Micro replacement purification column, activated Pure Solv media

required but not provided

Product #

Description

Comments

Add to Cart

Z560723 Micromatic MacroValve coupler for returnable containers, with 4F key Pure-Pac® II containers require the Micromatic Macrovalve for dispensing solvents.

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?