459844 Sigma-Aldrich

Ethanol

ACS reagent, ≥99.5% (200 proof), absolute

DOWNLOAD MSDS (PDF)

Synonym: Ethyl alcohol

Properties

Related Categories ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles, Ethanol (Tax-Paid Ethanol),
Quality Level   PREMIUM
grade   ACS reagent
vapor density   1.59 (vs air)
vapor pressure   44.6 mmHg ( 20 °C)
assay   ≥99.5% (200 proof)
autoignition temp.   683 °F
expl. lim.   19 %, 60 °F
impurities   H2SO4, passes test(darkened)
  MnO4- reducers, passes test
  acetone, passes test
  isopropanol, passes test
  ≤0.0002 meq/g Titr. base
  ≤0.0005 meq/g Titr. acid
  ≤0.1% Methanol
  ≤0.2% water
evapn. residue   ≤0.001%
color   APHA: ≤10
refractive index   n20/D 1.3600(lit.)
bp   78 °C(lit.)
  78.3 °C
mp   -114 °C
  −114 °C(lit.)
solubility   H2O: passes test
density   0.789 g/mL at 20 °C
  0.789 g/mL at 25 °C(lit.)

Description

Frequently Asked Questions

Live Chat and Frequently Asked Questions are available for this Product.

General description

Chromatin isolation by RNA purification (ChIRP). Ci C Chu, Jeffrey J Quinn, Howard Y HY Chang, Journal of Visualized Experiments 2012-01-01
Chromatin isolation by RNA purification (ChIRP)

Genomic maps of long noncoding RNA occupancy reveal principles of RNA-chromatin interactions.
Ci C Chu, Kun K Qu, Franklin L FL Zhong, Steven E SE Artandi, Howard Y HY Chang, Molecular Cell 2011-11-18Genomic maps of long noncoding RNA occupancy reveal principles of RNA-chromatin interactions.

Prices include Federal Tax.

Other Notes

Sales restrictions may apply

Packaging

1, 6×1, 2, 2.5, 4, 4×4 L in glass bottle

25 L in ss drum

200 L in steel drum

500, 6×500 mL in glass bottle

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Price and Availability

Customers Also Viewed

Acetone
650501Acetone

CHROMASOLV® Plus, for HPLC, ≥99.9%

Ethanol
E7023Ethanol

200 proof (absolute), for molecular biology

Ethanol
459836Ethanol

200 proof, anhydrous, ≥99.5%

Ethanol
493546Ethanol

200 proof, meets USP testing specifications

Ethanol
E7148Ethanol

for molecular biology, 190 proof

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?