• USA Home
  • 46008 - (1S,2S,5R)-(+)-Neomenthyl acetate

EMAIL THIS PAGE TO A FRIEND
46008 Sigma-Aldrich

(1S,2S,5R)-(+)-Neomenthyl acetate

puriss. p.a., terpene standard for GC, ≥99.0% (sum of enantiomers, GC)

  • CAS Number 2552-91-2

  • Empirical Formula (Hill Notation) C12H22O2

  • Molecular Weight 198.30

  •  Beilstein Registry Number 2208504

  •  EC Number 219-856-3

  •  MDL number MFCD00062978

Purchase

Properties

grade   terpene standard for GC
InChI Key   XHXUANMFYXWVNG-USWWRNFRSA-N
grade   puriss. p.a.
assay   ≥99.0% (sum of enantiomers, GC)
form   neat
optical activity   [α]20/D +48.0±1°, c = 8% in benzene
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   36-38 °C
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1325 4.1/PG 2
WGK Germany 
3
Flash Point(F) 
90 °F
Flash Point(C) 
32 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?