• USA Home
  • 46025 - Androstanolone 17-benzoate

EMAIL THIS PAGE TO A FRIEND
46025 Sigma-Aldrich

Androstanolone 17-benzoate

VETRANAL, analytical standard

Synonym: 17β-Hydroxy-5α-androstan-3-one 17-benzoate, 5α-Androstan-17β-ol-3-one benzoate, Dihydrotestosterone benzoate, Stanolone benzoate

  • CAS Number 1057-07-4

  • Empirical Formula (Hill Notation) C26H34O3

  • Molecular Weight 394.55

  •  Beilstein Registry Number 3122672

  •  EC Number 213-891-8

  •  MDL number MFCD00003666

  •  PubChem Substance ID 329757061

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Biomarkers and Metabolomics Standards,
grade   analytical standard
InChI Key   ZGDZDAPCWHIIKB-LVYWIKMTSA-N
product line   VETRANAL
form   neat
shelf life   limited shelf life, expiry date on the label
drug control   USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BV8058500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?