• USA Home
  • 46042 - 4-(Trifluoromethyl)umbelliferyl acetate

EMAIL THIS PAGE TO A FRIEND
46042 Sigma-Aldrich

4-(Trifluoromethyl)umbelliferyl acetate

for fluorescence, ≥98.0% (TLC)

  • CAS Number 141573-62-8

  • Empirical Formula (Hill Notation) C12H7F3O4

  • Molecular Weight 272.18

  •  MDL number MFCD00132979

Purchase

Properties

Related Categories Biochemicals and Reagents, Detection, Fluorescent Probes, Labels, Particles and Stains More...
grade   for fluorescence
InChI Key   JOHYKNHCOXRLMX-UHFFFAOYSA-N
assay   ≥98.0% (TLC)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   106-108 °C(lit.)
solubility   DMF: soluble
  DMSO: soluble
fluorescence   λex 410 nm; λem 503 nm in 0.1 M Tris pH 8.0 (esterase)
storage temp.   −20°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?