• USA Home
  • 46054 - Esterase from Bacillus thermoglucosidasius

EMAIL THIS PAGE TO A FRIEND
46054 Sigma-Aldrich

Esterase from Bacillus thermoglucosidasius

lyophilized, powder, beige, ~0.1 U/mg

Purchase

Description

Biochem/physiol Actions

Esterases are lipolytic enzymes that hydrolyze soluble fatty acid esters.

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol ethyl valerate (Cat. No. 30784) per minute at pH 8.0 and 25 °C

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?