EMAIL THIS PAGE TO A FRIEND
46101 Sigma-Aldrich

Ethyl cellulose

viscosity 80-170 mPa.s, 5 % in toluene/ethanol 80:20(25 °C)

Purchase

Properties

assay   48.0-49.5% (W/W)
form   coarse powder
autoignition temp.   698 °F
composition   ethoxyl content, 48.0-49.5 % (w/w)
refractive index   n20/D 1.47(lit.)
viscosity   80-170 mPa.s, 5 % in toluene/ethanol 80:20(25 °C)
density   1.14 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
FJ5950500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?