EMAIL THIS PAGE TO A FRIEND
46129 Sigma-Aldrich

Eugenol

tested according to Ph.Eur.

Synonym: 2-Methoxy-4-(2-propenyl)phenol, 4-Allyl-2-methoxyphenol, 4-Allylguaiacol, Eugenolum

  • CAS Number 97-53-0

  • Linear Formula 4-(H2C=CHCH2)C6H3-2-(OCH3)OH

  • Molecular Weight 164.20

  •  Beilstein Registry Number 1366759

  •  EC Number 202-589-1

  •  MDL number MFCD00008654

  •  PubChem Substance ID 24869841

Purchase

Properties

InChI Key   RRAFCDWBNXTKKO-UHFFFAOYSA-N
form   liquid
pharmacopeia   testing conforms to Pharmacopeia
refractive index   n20/D 1.541(lit.)
bp   254 °C(lit.)
mp   −12-−10 °C(lit.)
density   1.067 g/mL at 25 °C(lit.)
Gene Information   human ... UGT1A4(54657)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
SJ4375000
Flash Point(F) 
233.6 °F
Flash Point(C) 
112 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?