EMAIL THIS PAGE TO A FRIEND
46206 Sigma-Aldrich

3-Hexanone

analytical standard

Synonym: Ethyl propyl ketone

  • CAS Number 589-38-8

  • Linear Formula CH3CH2CH2COC2H5

  • Molecular Weight 100.16

  •  Beilstein Registry Number 1738025

  •  EC Number 209-645-4

  •  MDL number MFCD00009402

  •  PubChem Substance ID 329757100

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
InChI Key   PFCHFHIRKBAQGU-UHFFFAOYSA-N
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.400(lit.)
bp   123 °C(lit.)
density   0.815 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1224 - class 3 - PG 3 - Ketones, liquid, n.o.s., HI: all
WGK Germany 
3
RTECS 
MP1575000
Flash Point(F) 
73.4 °F
Flash Point(C) 
23 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?