• USA Home
  • 46254 - 1,1,1,2-Tetrachloroethane

EMAIL THIS PAGE TO A FRIEND
46254 Sigma-Aldrich

1,1,1,2-Tetrachloroethane

analytical standard

  • CAS Number 630-20-6

  • Linear Formula ClCH2CCl3

  • Molecular Weight 167.85

  •  Beilstein Registry Number 1733216

  •  EC Number 211-135-1

  •  MDL number MFCD00000807

  •  PubChem Substance ID 24869982

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
InChI Key   QVLAWKAXOMEXPM-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.481(lit.)
bp   138 °C(lit.)
density   1.598 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
2
RTECS 
KI8450000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?