• USA Home
  • 46269 - 1-(Ferrocenyl)ethanol

EMAIL THIS PAGE TO A FRIEND
46269 Sigma-Aldrich

1-(Ferrocenyl)ethanol

purum, ≥98.0% (HPLC)

Synonym: α-Ferrocenylethanol, α-Methylferrocenemethanol, (1-Hydroxyethyl)ferrocene, NSC 162110

  • CAS Number 1277-49-2

  • Empirical Formula (Hill Notation) C12H14FeO

  • Molecular Weight 230.08

  •  MDL number MFCD00013752

Purchase

Properties

InChI Key   NJPGTUIMEAOISI-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   76-79 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

• Ligand in titanium-catalyzed intermolecular hydroamination of terminal alkynes (Markovnikov vs. anti-Markovnikov addition)

Reactant for:
• Single-electron transfer living radical polymerization reactions
• Preparation of ferrocene-modified thiopyrimidines as anticancer agents
• Preparation of rersible addition-fragmentation chain transfer (RAFT) agent for RAFT polymerization of styrene

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
LK0730000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

369268 α-Methylferrocenemethanol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?