• USA Home
  • 46376 - Musk moskene solution

EMAIL THIS PAGE TO A FRIEND
46376 Sigma-Aldrich

Musk moskene solution

100 μg/mL in acetonitrile, analytical standard

Synonym: 1,1,3,3,5-Pentamethyl-4,6-dinitroindane

  • CAS Number 116-66-5

  • Empirical Formula (Hill Notation) C14H18N2O4

  • Molecular Weight 278.30

  •  Beilstein Registry Number 2163411

  •  EC Number 200-835-2

  •  MDL number MFCD00021242

  •  PubChem Substance ID 24870093

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Aromatics, Chemical Class,
grade   analytical standard
InChI Key   UHWURQRPEIFIAK-UHFFFAOYSA-N
concentration   100 μg/mL in acetonitrile
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?