• USA Home
  • 46381 - Florisil® Adsorbent for Chromatography

EMAIL THIS PAGE TO A FRIEND
46381 Sigma-Aldrich

Florisil® Adsorbent for Chromatography

16-30 mesh

Synonym: Activated magnesium silicate, Florisil®, Magnesium silicate

Purchase

Properties

Related Categories Adsorbents (Inorganic), Analytical/Chromatography, Florisil, LPLC Separation Media, LPLC/TLC/Paper Chromatography More...
grade   standard
InChI Key   FKHIFSZMMVMEQY-UHFFFAOYSA-N
form   granular
particle size   16-30 mesh
  595-1190 μm
surface area   289 m2/g

Description

Application

Florisil was used:
• as column packing material in column chromatography
• as an adsorbent in gas chromatography with electron capture detector (GC/ECD), capillary column chromatography, adsorption column chromatography
• in clean-up procedures in gas chromatography-tandem mass spectrometry and GC-FID

General description

Florisil, adsorbent for the separation of lipids, is a commercially-prepared magnesia silica gel. It has a coarse mesh size which helps in rapid flow rate. Florisil is effective in promoting [1,3]-sigmatropic shift of myceophenolic acid related allyl phenyl ethers. Florisil cartridges are used in atmospheric sampling to assess possible interference from air contaminants.

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?