EMAIL THIS PAGE TO A FRIEND
46387 Sigma-Aldrich

Florisil® TLC

for thin layer chromatography

Synonym: Magnesium silicate, Silica without Fluorescence

Purchase

Properties

Related Categories Adsorbents for TLC, Analytical/Chromatography, Florisil (Magnesium Silicate) TLC Adsorbent, LPLC/TLC/Paper Chromatography, Thin Layer Chromatography (TLC) More...
grade   for thin layer chromatography
InChI Key   FKHIFSZMMVMEQY-UHFFFAOYSA-N

Description

Application

Florisil® TLC was used in TLC separations of organic compounds.

General description

Florisil, adsorbent for the separation of lipids is a commercially-prepared magnesia silica gel. It has a coarse mesh size which helps in rapid flow rate. Florisil is effective in promoting [1,3]-sigmatropic shift of myceophenolic acid related allyl phenyl ethers. Florisil cartridges are used in atmospheric sampling to assess possible interference from air contaminants.

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?