• USA Home
  • 46442 - Dinoterb-bis(2-hydroxyethyl)ammonium solution

EMAIL THIS PAGE TO A FRIEND
46442 Sigma-Aldrich

Dinoterb-bis(2-hydroxyethyl)ammonium solution

100 ng/μL in acetonitrile, PESTANAL®, analytical standard

  • CAS Number 220045-55-6

  • Empirical Formula (Hill Notation) C10H12N2O5 · C4H11NO2

  • Molecular Weight 345.35

  •  MDL number MFCD00144151

  •  PubChem Substance ID 329757186

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, D, Dinitrophenol,
grade   analytical standard
InChI Key   MIINUZNZTMPLTJ-UHFFFAOYSA-N
product line   PESTANAL®
shelf life   limited shelf life, expiry date on the label
concentration   100 ng/μL in acetonitrile
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1648 3/PG 2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?