• USA Home
  • 46465 - 1-Adamantyl fluoroformate

EMAIL THIS PAGE TO A FRIEND
46465 Sigma-Aldrich

1-Adamantyl fluoroformate

technical, ≥90% (T)

  • CAS Number 62087-82-5

  • Empirical Formula (Hill Notation) C11H15FO2

  • Molecular Weight 198.23

  •  Beilstein Registry Number 3590131

  •  EC Number 263-400-6

  •  MDL number MFCD00074772

  •  PubChem Substance ID 24870203

Purchase

Properties

Related Categories Amine Protection, Chemical Biology, Chemical Synthesis, Other Protecting and Derivatizing Reagents, Peptide Chemistry,
InChI Key   PNLZKVOKOVFGMZ-KJZNFTALSA-N
grade   technical
assay   ≥90% (T)
mp   30-32 °C(lit.)
  30-32 °C
storage temp.   2-8°C

Description

Other Notes

Reagent suited for the protection of imidazole function of histidine; Used for the protection of ω-amino groups of Lys and Orn; e.g. in fragment condensations; γ,ω-bis-protection of arginine

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
Flash Point(F) 
190.4 °F
Flash Point(C) 
88 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?